Order Entry
Finland
ContactUsLinkComponent
Beakers, low form, stainless steel
Beakers, low form, stainless steel
Catalog # AGEN2300A
Supplier:  Agencinox
Supplier #: 2300A
undefined
Beakers, low form, stainless steel
Catalog # AGEN2300A
Supplier:  Agencinox
Supplier #: 2300A
Log in for availability
Select an Option
150 mm
Select an Option
3000 ml
Select an Option
180 mm
Select an Option
17
Select an Option
1
About this item
Stainless steel.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.