Capillary columns, TRACE™ TR-35MS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Mid-polarity columns excellent for many applications.
- Mid-polarity phase, 35% phenyl polysilphenylene-siloxane
- Exceptionally low surface activity
- Low bleed even at elevated temperatures