96-Well assay plates, 'V' well, Axygen®
Supplier: Corning
Certificates
About this item
Axygen® assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
- SBS footprint
- Ultra-flat surface ensures proper sealing with heat sealing film
- Certified free of RNase, DNase and pyrogens
Specifications
- Catalog No:
- AXYGP-96-450V-B
- AXYGP-96-450V-Y
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa