Storage cabinets for acids and alkalis, SLS 600
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
These storage cabinet body made from HPL-coated sheets, particularly impact, scratch and abrasion proof are used for storage of acids and alkalis.
- Doors with separate cylinder locks
- Levelling feet for installing cabinet horizontally
- Supply air by ventilation grille in the back wall
- Colour RAL 7035 - light grey
- Ventilation connection for exhaust air extraction on cabinet roof
Inner body and inner fittings completely made from corrosion-proof components.
Cabinet features with central horizontal division for storage of acids and alkalis in separate compartments.
Delivery: Supplied with full extent pull-out drawers with built-in PE sump tray (20 l).