Order Entry
Finland
ContactUsLinkComponent
Dodecamolybdophosphoric acid solution in 2-propanol, spray reagent for TLC, Supelco®
Flammable
Corrosive
Irritant
Dodecamolybdophosphoric acid solution in 2-propanol, spray reagent for TLC, Supelco®
Catalog # 1.00480.0100
Supplier:  Merck
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Dodecamolybdophosphoric acid solution in 2-propanol, spray reagent for TLC, Supelco®
Catalog # 1.00480.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :1825.25 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ100
UN:1219
 :GHS02
 :GHS05
 :GHS07
CAS Number:12026-57-2
Formula:H₃Mo₁₂O₄₀P
MDL Number:MFCD00011341