
5-Bromophthalide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64169-34-2
:
Formula:C₈H₅BrO₂
MDL Number:MFCD01797360
:213.03 g/mol
Melting Pt:162…166 °C
EINECS:447-190-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:162 to 166 °C
- HPLC:≥97.5 %