Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethyl-3-hexanone
2,2-Dimethyl-3-hexanone ≥99%
2,2-Dimethyl-3-hexanone ≥99%
Irritant
2,2-Dimethyl-3-hexanone ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2,2-Dimethyl-3-hexanone ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
5405-79-8
:
Formula:
C₈H₁₆O
MDL Number:
MFCD00027140
:
128.21 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB20612
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation