Direct red 80
TCIAD3273-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Red to Dark blue to Black powder to crystal
- Lambda max.:524.0 to 528.0 nm (H2O)
- Absorbance (E1%1cm):min. 350 (H2O, 524.0 to 528.0 nm)
:
:1373.051 g/mol
:14,08548
:220-027-3
:2610-10-8
:C₄₅H₂₆N₁₀Na₆O₂₁S₆
:MFCD00054389