Order Entry
Finland
Orders LinkContactUsLinkComponent
Slow worm model

Slow worm model

Slow worm model, SOMSO-Plast®, 1,1 kg, 180×180×340 mm
Catalog # SOMMZOS1026
Supplier:   SOMMER MARCUS
Slow worm model
Catalog # SOMMZOS1026
Supplier:  SOMMER MARCUS
Slow worm model
Slow worm model

Slow worm model, SOMSO-Plast®, 1,1 kg, 180×180×340 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Anguis fragilis.

Adolescent worm with printed label and description.

Packaging: In a transparent box.

Specifications
  • Weight:
    1,1 kg
  • Description:
    Slow worm model
  • Material:
    SOMSO-Plast®
  • W×D×H:
    180×180×340 mm