Order Entry
Finland
ContactUsLinkComponent
Common female toad model
Common female toad model
Catalog # SOMMZOS1013
Supplier:  SOMMER MARCUS
undefined
Common female toad model
Catalog # SOMMZOS1013
Supplier:  SOMMER MARCUS

Specifications

  • Weight:
    1,1 kg
  • Description:
    Common female toad
  • Material:
    SOMSO-Plast®
  • W×D×H:
    180×180×340 mm
  • Pk:
    1

Specifications

About this item

Bufo bufo.

Includes a printed label and description.

Packaging: In a transparent box.