Order Entry
Finland
ContactUsLinkComponent
 
Functional model of the larynx
Catalog # SOMMGS10
Supplier:  SOMMER MARCUS
undefined
Functional model of the larynx
Catalog # SOMMGS10
Supplier:  SOMMER MARCUS

Specifications

  • Weight:
    1,5 kg
  • Description:
    Functional model of the larynx
  • Material:
    SOMSO-Plast®
  • W×D×H:
    320×180×180 mm
  • Magnification:
  • Pk:
    1

Specifications

About this item

The opening and closing of the true glottis, the variation of tension of the vocal fold and the passage of air can be demonstrated very instructively.

Packaging: On a green base.