APA6000 Alkalinity Reagent 1
Catalog # HACH2696553
Supplier: Hach
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:APA6000 Alkalinity Reagent 1
- Storage temperature:Ambient
Specifications
About this item
Caution: Hazard components for labelling sulphuric acid 1 %, Corrosive