0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Methyl orange
Methyl orange 0.1% in ethanol 20% (v/v)
Methyl orange 0.1% in ethanol 20% (v/v)
REGEMTR05025
:
REAGECON
:
Methyl orange 0.1% in ethanol 20% (v/v)
REGEMTR05025
:
REAGECON
:
Pack type:
Glass bottle
Pk:
250 mL
Seq:
2590
:
:
327.34 g/mol
:
208-925-3
:
1170
:
3,III
:
547-58-0
:
C₁₄H₁₄N₃NaO₃S
:
MFCD00007502