Methyl orange 0.1% in ethanol 20% (v/v) Reag. Ph. Eur.
Catalog # FFCHFF0930500
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:327.34 g/mol
EINECS:208-925-3
UN:1170
ADR:3,III
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502