Ammonium chloride 1 mol/l (1 N) in aqueous solution
Catalog # FFCHFF63919020
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:20 L
- Seq:5302
Specifications
About this item
:
:53.49 g/mol
Storage Temperature:Ambient
EINECS:235-186-4
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420