Bromothymol blue 0.04% in water for pH determination
Catalog # TCIAB0658-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Transition interval (pH):(yellow)6.0 to 7.6 (blue)
- pH:5.0 to 7.0
Specification Test Results
About this item
:
Merck Index:14,01445
:624.39 g/mol
Storage Temperature:Ambient
EINECS:200-971-2
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872