Order Entry
Finland
ContactUsLinkComponent
Methyl orange 0.1% in water for titration
Methyl orange 0.1% in water for titration
Catalog # TCIAM0017-500ML
Supplier:  TCI
CAS Number:  
undefined
Methyl orange 0.1% in water for titration
Catalog # TCIAM0017-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Red to Dark red to Brown clear liquid
  • Transition interval (pH):
    (yellowish red)3.1 to 4.4 (redish yellow)

Specification Test Results

About this item

 :
 :327.34 g/mol
Merck Index:14,06105
Storage Temperature:Ambient
EINECS:208-925-3
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502