Order Entry
Finland
ContactUsLinkComponent
Ammonium chloride 0.1 g/l in aqueous solution
Ammonium chloride 0.1 g/l in aqueous solution
Catalog # FFCHFF9220500
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Ammonium chloride 0.1 g/l in aqueous solution
Catalog # FFCHFF9220500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :53.49 g/mol
Storage Temperature:Ambient
EINECS:235-186-4
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420