Order Entry
Finland
ContactUsLinkComponent
3-Phenoxypropiononitrile ≥98.0% (by GC)
Irritant
3-Phenoxypropiononitrile ≥98.0% (by GC)
Catalog # TCIAP1426-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-Phenoxypropiononitrile ≥98.0% (by GC)
Catalog # TCIAP1426-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    59.0 to 63.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
Boiling Pt:120°C
EINECS:221-278-1
 :GHS07
CAS Number:3055-86-5
Formula:C₉H₉NO
MDL Number:MFCD00013822