Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-3-methoxyquinoline
5-Bromo-3-methoxyquinoline 96%
5-Bromo-3-methoxyquinoline 96%
Irritant
5-Bromo-3-methoxyquinoline 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
5-Bromo-3-methoxyquinoline 96%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
776296-12-9
:
Formula:
C₁₀H₈BrNO
MDL Number:
MFCD16038652
:
238.08 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAH54919
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation