3-Methoxy-5-quinolineboronic acid pinacol ester 96%
Catalog # H54211.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:881656-35-5
:
Formula:C₁₆H₂₀BNO₃
MDL Number:MFCD20527212
:285.15 g/mol
Storage Temperature:Ambient