Order Entry
Finland
Orders LinkContactUsLinkComponent
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%

3,5-Dimethyl-4-hydroxybenzonitrile ≥98%

Toxic
Irritant
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%
Certificates
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%
3,5-Dimethyl-4-hydroxybenzonitrile ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 4198-90-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 123-127° MDL No.: MFCD00221716

Synonyms:  4-Hydroxy-3,5-dimethylbenzonitrile
  • CAS Number:4198-90-7
  • Formula:C₉H₉NO
  • MDL Number:MFCD00221716
  • Molecular Weight:147.18 g/mol
  • Melting Pt:123…127 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information