
4-Cyano-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:843663-18-3
:
Formula:C₇H₅BFNO₂
MDL Number:MFCD03411549
:164.93 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.5 %