Order Entry
Finland
ContactUsLinkComponent
 
Bromothymol blue 1% in DMF Reag. Ph. Eur. 1012902
  87797.180
 :  
undefined
Bromothymol blue 1% in DMF Reag. Ph. Eur. 1012902
  87797.180
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

 :
 :152 °C
 :–61 °C
 :58 °C (closed cup)
 :3,III
 :624.39 g/mol
 :200-971-2
 :2265
 :GHS02
 :GHS08
 :GHS07
 :Ambient
 :76-59-5
 :C₂₇H₂₈Br₂O₅S
 :MFCD00005872