Sodium hydroxide 6.25 mol/l (6.25 N) in aqueous solution
Catalog # 85100.360
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:1.22 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548