Order Entry
Finland
ContactUsLinkComponent
Cut resistant gloves, SivaCut® 830
Cut resistant gloves, SivaCut® 830
Supplier:  KCL by Honeywell
Certificates
undefined
Cut resistant gloves, SivaCut® 830
Supplier:  KCL by Honeywell
Specifications
  • Catalog No:
  • KCHL830.11
  • KCHL830.10
  • KCHL830.7
  • KCHL830.8
  • KCHL830.9
  • Model:
  • 830
  • 830
  • 830
  • 830
  • 830
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Finish:
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Coating on palm and fingers
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388 (2543)
  • EN 388 (2543)
  • EN 388 (2543)
  • EN 388 (2543)
  • EN 388 (2543)
Specifications