Order Entry
Finland
ContactUsLinkComponent
Mussel model
Mussel model
Catalog # SOMMZOS119
Supplier:  SOMMER MARCUS
undefined
Mussel model
Catalog # SOMMZOS119
Supplier:  SOMMER MARCUS

Specifications

  • Weight:
    7,7 kg
  • Description:
    Mussel
  • Material:
    SOMSO-Plast®
  • W×D×H:
    610×380×210 mm
  • Pk:
    1

Specifications

About this item

Anodonta cygnea. Anatomical view, right half of shell - pallium half and gill removed, foot opened at the right side. The model shows the special features of the external and internal structure of a mussel. The organ systems, which are to be seen only with difficulty on the natural preparation have been accentuated and schematised necessarily, only for didactic reasons in structure and colour.

  • Linearly enlarged four times
  • Made in SOMSO-Plast®
  • Model separates into five parts
Packaging: On a green base.