Crayfish model
Crayfish model, SOMSO-Plast®, 4 kg, 820×290×280 mm
Catalog # SOMMZOS118
Supplier: SOMMER MARCUS
Jump to:
Product Details & Documents
Astacus astacus (L.). Structure of the body and anatomy of a male animal.
- Linearly enlarged three times
- Model can separate into 13 parts
- Movable claws to demonstrate the position of the internal organs
The model becomes of special value for demonstration: The lifelike model shows on the left side the differentiated external limbs, on the right side the internal structure of the crayfish.
Packaging: On a stand with green base.
Specifications
- Weight:4 kg
- Description:Crayfish model
- Material:SOMSO-Plast®
- W×D×H:820×290×280 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



