Order Entry
Finland
Orders LinkContactUsLinkComponent
Star fish model

Star fish model

Star fish model, SOMSO-Plast®, 2,2 kg, 530×350×310 mm
Catalog # SOMMZOS114
Supplier:   SOMMER MARCUS
Star fish model
Catalog # SOMMZOS114
Supplier:  SOMMER MARCUS
Star fish model
Star fish model

Star fish model, SOMSO-Plast®, 2,2 kg, 530×350×310 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Asterias. The model shows:

  • 1. Complete arm with normal position of the organs (stomach, intestinal caeca, pyloric caica, ampullae)
  • 2. Arm with ampullae, ring canal, Polian vesicles
  • 3. Skeleton of the arm with nervous system (nerve-ring and radial nerves)
  • 4. Arm in transverse section (ambulacral system in cross-section)

The cut surface shows the stomach, stone canal, axial organ and madreporite. In addition on the outside the podia, pedicellariae and optic organs are shown.

Packaging: On a removable stand and a green base.

Specifications
  • Weight:
    2,2 kg
  • Description:
    Star fish model
  • Material:
    SOMSO-Plast®
  • W×D×H:
    530×350×310 mm