Single cell changing animalcule model
SOMMZOS101
- Weight:1,8 kg
- Description:Single cell changing animalcule model
- Material:SOMSO-Plast®
- W×D×H:480×310×80 mm
- Pk:1
Amoeba proteus.
- Enlarged around 1000 times linearly
- Adapted to modern standards of research
- Model separates into two parts
In a small pseudo-podium which can be opened up showing the structure after electronmicroscopic magnification.
Weight: 1,8 kg
: On a green base with explanatory notes.