Order Entry
Finland
Orders LinkContactUsLinkComponent
Bony fish model

Bony fish model

Model of a bony fish, SOMSO-Plast®, 1,6 kg, 490×150×350 mm
Catalog # SOMMZOS105
Supplier:   SOMMER MARCUS
Bony fish model
Catalog # SOMMZOS105
Supplier:  SOMMER MARCUS
Bony fish model
Bony fish model

Model of a bony fish, SOMSO-Plast®, 1,6 kg, 490×150×350 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Cyprinus carpio. Carp model.

  • Modelled from life in natural size
  • Intestines, bladder and testicles are removable
  • Model separates into four parts

Packaging: On a stand with green base and explanation.

Specifications
  • Weight:
    1,6 kg
  • Description:
    Model of a bony fish
  • Material:
    SOMSO-Plast®
  • W×D×H:
    490×150×350 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.