Functional elbow model
Functional model of the elbow, SOMSO-Plast®, 0,65 kg, 410×190×220 mm
Catalog # SOMMNS52
Supplier: SOMMER MARCUS
Jump to:
Product Details & Documents
The following movements are possible:
- Extension and flexion of the upper arm and forearm
- Rotation movement of the radius
- Pronation and supination
: Removable from stand with green base.
Specifications
- Weight:0,65 kg
- Description:Functional model of the elbow
- Material:SOMSO-Plast®
- W×D×H:410×190×220 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



