Order Entry
Finland
Orders LinkContactUsLinkComponent
Brain model

Brain model

Brain in two parts, SOMSO-Plast®, 800 g, 160×150×170 mm
Catalog # SOMMBS21
Supplier:   SOMMER MARCUS
Brain model
Catalog # SOMMBS21
Supplier:  SOMMER MARCUS
Brain model
Brain model

Brain in two parts, SOMSO-Plast®, 800 g, 160×150×170 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Brain model in two parts.

  • Median section
  • Natural cast on a base
Specifications
  • Weight:
    800 g
  • Description:
    Brain in two parts
  • Material:
    SOMSO-Plast®
  • W×D×H:
    160×150×170 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.