Order Entry
Finland
Orders LinkContactUsLinkComponent
Lymphatic system

Lymphatic system

Lymphatic system, 10 kg, 840×540×120 mm
Catalog # SOMMHS19/1
Supplier:   SOMMER MARCUS
Lymphatic system
Catalog # SOMMHS19/1
Supplier:  SOMMER MARCUS
Lymphatic system
Lymphatic system

Lymphatic system, 10 kg, 840×540×120 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Relief model of approximately 2/3 natural size in one piece.

 : On a green base plate.

Specifications
  • Weight:
    10 kg
  • Description:
    Lymphatic system
  • W×D×H:
    840×540×120 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.