Safety shoes, lace-up, Steelite™ Executive Brogue FW46
Supplier: Portwest
Certificates
About this item
These laced executive brogue office/service shoes are manufactured from Brazilian full grain leather plus an oil resistant, dual-density PU/PU outer sole.
- Special moisture management fabric for extra comfort
- Shock-absorbent heel
- Protective steel toecap (200 J)
- Puncture resistant steel midsole
Colour: Black
Certifications: EN ISO 20345:2011; S1P, SRC.