Conical tubing adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
PEEK.
- Direct connect ¹/₁₆ and ¹/₈" ext. Ø rigid and semi-rigid tubing to peristaltic tubing
- Accept 0,020 to ¹/₈" (0,50 to 3,2 mm) int. Ø peristaltic tubing
- Biocompatible flowpath with excellent chemical compatibility
Upchurch Scientific® Conical adapters provide a reliable connection between rigid/semi-rigid tubing and peristaltic-type flexible tubing, such as Tygon® and PharMed®. These adapters are composed of a PEEK polymer female nut, our Super Flangeless™ ferrule system and a Tefzel® (ETFE) or PEEK conical adapter body. The narrow coned end of the adapter body allows peristaltic tubing to slide on more easily than it does onto conventional barbed adapters. Peristaltic tubing is also easier to remove from conical adapters, since no cutting or excessive pulling is required.