Intensifying screens, BioMax® MS
Supplier: CARESTREAM HEALTH
Certificates
About this item
The BioMax® MS intensifying screen is optimised for use with BioMax® MS film to maximise sensitivity for detection of ³²P and ¹²⁵I labelled samples.
The BioMax® MS intensifying screen may be used with any autoradiography film to increase sensitivity. However, maximum sensitivity is achieved in combination with the BioMax® MS film.