Intensifying screens, BioMax® TranScreen HE
Supplier: CARESTREAM HEALTH
Certificates
About this item
Designed for use with gamma emitters such as ¹²⁵I and high-energy beta emitters, such as ³²P, the BioMax™ TranScreen HE intensifying screen offers the greatest sensitivity. When used in conjunction with the BioMax™ MS film, the system equals or exceeds the image quality of conventional autoradiography screen/film systems. By minimising the distances between the radioisotope and phosphor layer, it presents the phosphor to the emitting particles immediately, for less scattering and better spatial resolution.