Capillary columns, TraceGOLD™ TG-VMS
Supplier: Thermo Fisher Scientific
Certificates
About this item
TraceGOLD™ TG-VMS GC columns are ideal for volatile organic pollutants.
- Highly stable polymer phase
- Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity