
Methyl-2,3-dihydroxybenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2411-83-8
:
Formula:C₈H₈O₄
MDL Number:MFCD00039520
:168.1467 g/mol
Boiling Pt:96°C
EINECS:219-317-2
:GHS07
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:78.0 to 82.0 deg-C