Order Entry
Finland
ContactUsLinkComponent
Infusion valve, Safeflow®
Infusion valve, Safeflow®
  BRAU409100H
 :  B. Braun
undefined
Infusion valve, Safeflow®
  BRAU409100H
 :  B. Braun
  • Flow rate:
    360 ml/min
  • Volume:
    0,9 ml
  • Colour:
    Purple/blue
  • Description:
    Infusion valves
  • Pk:
    50
This capless valve allows for a safe and convenient access during infusion therapy.
  • Rapid delivery
  • Compatible with luer-slip and -lock
  • Provides needle-free IV access while eliminating the risk of needle stick injuries
Suitable for every IV procedure.