
Dimethyl-trans-cyclohexane-1,4-dicarboxylate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3399-22-2
:
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00063917
Boiling Pt:140°C
:200.23 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:67.0 to 71.0 deg-C
Specification Test Results
Frequently Bought Together