
4-(Methylsulphonyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:149104-88-1
:
Formula:C₇H₉BO₄S
MDL Number:MFCD01630820
:200.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):97.0 to 110.0 %