Order Entry
Finland
ContactUsLinkComponent
 
 
1H-Pyrazole-4-boronic acid pinacol ester ≥97.0% (by GC)
 :  TCI
 
undefined
 
1H-Pyrazole-4-boronic acid pinacol ester ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    145.0 to 150.0 deg-C