Histamine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56-92-8
:
Formula:C₅H₁₁Cl₂N₃
MDL Number:MFCD00012703
:184.07 g/mol
Merck Index:14,04719
Melting Pt:246°C
EINECS:200-298-4
:GHS07
:GHS08
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency