

Histamine dihydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-92-8
:
Formula:C₅H₁₁Cl₂N₃
MDL Number:MFCD00012703
:184.07 g/mol
Merck Index:14,4719
Melting Pt:245…249 °C
EINECS:200-298-4
:GHS08
:GHS07
Storage Temperature:Ambient