Capillary columns, TraceGOLD™ TG-200MS
Supplier: Thermo Fisher Scientific
Certificates
About this item
TraceGOLD™ TG-200MS GC columns are exceptionally inert mid-polarity columns with selectivity and elution order optimised for difficult separations.
- Polar phase, Trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
- Outstanding thermal stability and low bleed
- Suitable for use with sensitive detectors including ECDs, NPDs and MS
- Equivalent to USP G6 phase
- Confirmation column in combination with another GC column