Superficially porous particle (SPP) columns, Brownlee™ C8
Supplier: PerkinElmer
Certificates
About this item
These superficially porous particle (SPP) columns produce sharper peaks and faster separation results due to their breakthrough particle design and size.
- General purpose octyl phase for reversed phase separations with less retension than C18
- Exhibits peak shape for wide range of compounds
- pH range of 2 to 9 and the temperature range of 60 °C
The design allows for a shorter diffusion path, reducing the time solute molecules spend inside the particles while passing through the stationary phase.