Supelco® Organic Reference Standard, Scopoletin
Catalog # 38332-10MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:92-61-5
- Description:Scopoletin
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00006872
- M-Clarity quality level:MQ100
- Synonym(s):6-Methoxyumbelliferone; 6-Methylesculetin; 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one; 7-Hydroxy-6-methoxycoumarin; Buxuletin; Chrysatropic acid; Escopoletin; Esculetin-6-methyl ether; Gelseminic acid; Murrayetin
- Melting point:203...205 °C (lit.)
- Molecular weight:192,17
- Flash point:Not applicable