Melting point standard, vanillin, pharmaceutical secondary standard, Supelco®
Catalog # PHR1091-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Description:Vanillin
- Melting pt.:∼82 °C
Specifications
About this item
Certifications: This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470). Traceable to USP 1711009